N-Benzylglycine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl

B3075-25G B3075-5G
TCB3075-25GEA 182.43 CAD
TCB3075-25G TCB3075-5G
N-Benzylglycine hydrochloride ≥98.0% (by HPLC)
N-Benzylglycine hydrochloride

CAS Number: 7689-50-1
MDL Number: MFCD00156934
Molecular Formula: C9H11NO2
Molecular Weight: 201.65
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 228


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Bzl-Gly-OH.HCl

Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
Melting Pt: 228 °C
MDL Number: MFCD00156934
CAS Number: 7689-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbdd7f9bd8e5fa3ddaf417f8fd67f84bfafb8449&vid=dbdd7f9bd8e5fa3ddaf417f8fd67f84bfafb8449&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR