4-Bromo-3-nitrobenzotrifluoride ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromo-3-nitrobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-3-nitrotoluene, 1-Bromo-2-nitro-4-(trifluoromethyl)benzene

B3074-5G B3074-25G
TCB3074-5GEA 109.02 CAD
TCB3074-5G TCB3074-25G
4-Bromo-3-nitrobenzotrifluoride ≥98.0%
4-Bromo-3-nitrobenzotrifluoride

CAS Number: 349-03-1
MDL Number: MFCD00014685
Molecular Formula: C7H3BrF3NO2
Molecular Weight: 270.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 113
Specific Gravity (20/20): 1.80


Other Product Names / Synonyms: 1-Bromo-2-nitro-4-(trifluoromethyl)benzene

Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
Boiling Pt: 105 °C
Density: 1.80 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00014685
CAS Number: 349-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44c6025e74e98ddff0e50eefcdb071274562ad7b&vid=44c6025e74e98ddff0e50eefcdb071274562ad7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR