Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B3071-5G B3071-1G
TCB3071-5GEA 188.02 CAD
TCB3071-5G TCB3071-1G
Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine ≥98.0% (by HPLC, titration analysis)
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine

CAS Number: 92122-45-7
MDL Number: MFCD00065660
Molecular Formula: C26H32N2O6
Molecular Weight: 468.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: 2.2 deg (C=1, MeOH)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nepsilon-Boc-Nalpha-Fmoc-D-lysine, Fmoc-D-Lys(Boc)-OH

Formula: C₂₆H₃₂N₂O₆
Melting Pt: 130 °C
MDL Number: MFCD00065660
CAS Number: 92122-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df08e5df9a1894db1e8e7955c026280c584c7567&vid=df08e5df9a1894db1e8e7955c026280c584c7567&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR