3-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B3056-5G
B3056-25G
TCB3056-5GEA
118.5
CAD
TCB3056-5G
TCB3056-25G
3-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
3-(Benzyloxy)phenylboronic acid
CAS Number: 156682-54-1
MDL Number: MFCD02093063
Molecular Formula: C13H13BO3
Molecular Weight: 228.05
Form: Crystal
Color: White
Melting point (°C): 130
Other Product Names / Synonyms: 3-Benzyloxybenzeneboronic Acid
Formula:
C₁₃H₁₃BO₃ Melting Pt: 130 °C |
MDL Number:
MFCD02093063 CAS Number: 156682-54-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...