(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98.0%
Supplier: TCI America
B3054-1G
B3054-5G
TCB3054-1GEA
58.46
CAD
TCB3054-1G
TCB3054-5G
(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98.0%
(R)-(-)-1-Boc-3-hydroxypyrrolidine
CAS Number: 109431-87-0
MDL Number: MFCD01317838
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 61
Flash Point (°C): 110
Specific rotation [a]20/D: -27 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Boc-3-pyrrolidinol, (R)-1-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine, (R)-1-Boc-3-hydroxypyrrolidine
Formula:
C₉H₁₇NO₃ Melting Pt: 61 °C Flash Pt: 110 °C |
MDL Number:
MFCD01317838 CAS Number: 109431-87-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...