3-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3049-25G
B3049-5G
TCB3049-25GEA
123.24
CAD
TCB3049-25G
TCB3049-5G
3-Bromo-p-toluic acid ≥98.0% (by GC, titration analysis)
3-Bromo-p-toluic acid
CAS Number: 7697-26-9
MDL Number: MFCD00002488
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Bromo-p-toluic Acid
Formula:
C₈H₇BrO₂ Melting Pt: 210 °C |
MDL Number:
MFCD00002488 CAS Number: 7697-26-9 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...