1-Bromo-2-chloro-5-fluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-1-chloro-4-fluorobenzene

B3048-25G B3048-5G
TCB3048-25GEA 254.38 CAD
TCB3048-25G TCB3048-5G
1-Bromo-2-chloro-5-fluorobenzene ≥98.0%
1-Bromo-2-chloro-5-fluorobenzene

CAS Number: 201849-15-2
MDL Number: MFCD00061120
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.75

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Density: 1.75 (20 °C)
MDL Number: MFCD00061120
CAS Number: 201849-15-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ec7ec7595e651064147c194ce287d865bb08034&vid=7ec7ec7595e651064147c194ce287d865bb08034&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR