1-Bromo-3-chloro-5-fluorobenzene ≥98.0%

Supplier: TCI America

B3045-25G B3045-5G
TCB3045-25GEA 151.68 CAD
TCB3045-25G TCB3045-5G
1-Bromo-3-chloro-5-fluorobenzene ≥98.0%
1-Bromo-3-chloro-5-fluorobenzene

CAS Number: 33863-76-2
MDL Number: MFCD00070747
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Specific Gravity (20/20): 1.72

Formula: C₆H₃BrClF
Boiling Pt: 72 °C
Density: 1.72 (20 °C)
MDL Number: MFCD00070747
CAS Number: 33863-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b70d42a4178b0cd52ceb0b416817a7f3dccd709d&vid=b70d42a4178b0cd52ceb0b416817a7f3dccd709d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR