Bis(3-fluorophenyl) disulfide ≥97.0%
Supplier: TCI America
B3044-5G
B3044-1G
TCB3044-5GEA
116.92
CAD
TCB3044-5G
TCB3044-1G
Bis(3-fluorophenyl) disulfide ≥97.0%
Bis(3-fluorophenyl) disulphide
CAS Number: 63930-17-6
MDL Number: MFCD01318104
Molecular Formula: C12H8F2S2
Molecular Weight: 254.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.32
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,3'-Difluorodiphenyl Disulfide
Formula:
C₁₂H₈F₂S₂ Density: 1.32 (20 °C) |
MDL Number:
MFCD01318104 CAS Number: 63930-17-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...