2-Bromo-5-nitroaniline ≥97.0%

Supplier: TCI America

B3041-5G B3041-25G
TCB3041-5GEA 115.34 CAD
TCB3041-5G TCB3041-25G
2-Bromo-5-nitroaniline ≥97.0%
2-Bromo-5-nitroaniline

CAS Number: 10403-47-1
MDL Number: MFCD00051578
Molecular Formula: C6H5BrN2O2
Molecular Weight: 217.02
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 140

Formula: C₆H₅BrN₂O₂
Melting Pt: 140 °C
MDL Number: MFCD00051578
CAS Number: 10403-47-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a2df6a16f2c1891b96bc5dee0fa733bface4921&vid=80b3fc01b9de5a0b8708f95e5ae172f3ff0eb31e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR