6-Bromoquinaldine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B3039-5G
B3039-25G
TCB3039-5GEA
143.78
CAD
TCB3039-5G
TCB3039-25G
6-Bromoquinaldine ≥98.0% (by GC, titration analysis)
6-Bromoquinaldine
CAS Number: 877-42-9
MDL Number: MFCD00079724
Molecular Formula: C10H8BrN
Molecular Weight: 222.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 104
Other Product Names / Synonyms: 6-Bromoquinaldine
Formula:
C₁₀H₈BrN MW: 222.08 Melting Pt: 104 °C |
MDL Number:
MFCD00079724 CAS Number: 877-42-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...