4-Bromo-2-nitroaniline ≥98.0%

Supplier: TCI America

B3033-25G B3033-5G
TCB3033-25GEA 191.18 CAD
TCB3033-25G TCB3033-5G
4-Bromo-2-nitroaniline ≥98.0%
4-Bromo-2-nitroaniline

CAS Number: 875-51-4
MDL Number: MFCD00041312
Molecular Formula: C6H5BrN2O2
Molecular Weight: 217.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 112


Other Product Names / Synonyms: 1-Amino-4-bromo-2-nitrobenzene

Formula: C₆H₅BrN₂O₂
Melting Pt: 112 °C
MDL Number: MFCD00041312
CAS Number: 875-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19ae7fc104157bb674f63d213081edc89bcc9817&vid=19ae7fc104157bb674f63d213081edc89bcc9817&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR