2-Bromo-4-chlorotoluene ≥98.0%

Supplier: TCI America

Synonyms: 1-Bromo-3-chloro-6-methylbenzene, 2-Bromo-4-chloro-1-methylbenzene

B3031-25G B3031-5G
TCB3031-25GEA 126.4 CAD
TCB3031-25G TCB3031-5G
2-Bromo-4-chlorotoluene ≥98.0%
2-Bromo-4-chlorotoluene

CAS Number: 27139-97-5
MDL Number: MFCD00060650
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Specific Gravity (20/20): 1.56

Formula: C₇H₆BrCl
Boiling Pt: 114 °C
Density: 1.56 (20 °C)
MDL Number: MFCD00060650
CAS Number: 27139-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4aecc90dbbd066ce96a962578fc50a5a0d1c51ac&vid=4aecc90dbbd066ce96a962578fc50a5a0d1c51ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR