4-Bromo-2-chloro-1-iodobenzene ≥97.0%

Supplier: TCI America

Synonyms: 1-Bromo-3-chloro-4-iodobenzene

B3030-25G B3030-5G
TCB3030-25GEA 343.81 CAD
TCB3030-25G TCB3030-5G
4-Bromo-2-chloro-1-iodobenzene ≥97.0%
1-Bromo-3-chloro-4-iodobenzene

CAS Number: 31928-47-9
MDL Number: MFCD00079709
Molecular Formula: C6H3BrClI
Molecular Weight: 317.35
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 104
Freezing point (°C): 33

Formula: C₆H₃BrClI
MW: 190.45
Boiling Pt: 104 °C
MDL Number: MFCD00079709
CAS Number: 31928-47-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df3a8e9dadd560b5acb5c515254c696832351d30&vid=df3a8e9dadd560b5acb5c515254c696832351d30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR