6-Bromopicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B3024-5G B3024-1G
TCB3024-5GEA 201.61 CAD
TCB3024-5G TCB3024-1G
6-Bromopicolinic acid ≥98.0% (by GC, titration analysis)
6-Bromopicolinic acid

CAS Number: 21190-87-4
MDL Number: MFCD00093197
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 189


Other Product Names / Synonyms: 6-Bromopicolinic Acid

Formula: C₆H₄BrNO₂
Melting Pt: 189 °C
MDL Number: MFCD00093197
CAS Number: 21190-87-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aec54766468eb022a45719b26be3bf0cc89bb79c&vid=aec54766468eb022a45719b26be3bf0cc89bb79c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR