2,4-Bis(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

B3022-5G B3022-1G
TCB3022-5GEA 164.32 CAD
TCB3022-5G TCB3022-1G
2,4-Bis(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2,4-Bis(trifluoromethyl)phenylboronic acid

CAS Number: 153254-09-2
MDL Number: MFCD01631349
Molecular Formula: C8H5BF6O2
Molecular Weight: 257.93
Form: Crystal
Color: White


Other Product Names / Synonyms: 2,4-Bis(trifluoromethyl)benzeneboronic Acid

Formula: C₈H₅BF₆O₂
MW: 257.93 g/mol
MDL Number: MFCD01631349
CAS Number: 153254-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0aa987f97d864d48f40c9b8635eab1c70f30d2c&vid=b0aa987f97d864d48f40c9b8635eab1c70f30d2c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR