1-Bromo-2-fluoro-4-iodobenzene ≥97.0%

Supplier: TCI America

B3017-5G B3017-25G
TCB3017-5GEA 82.16 CAD
TCB3017-5G TCB3017-25G
1-Bromo-2-fluoro-4-iodobenzene ≥97.0%
1-Bromo-2-fluoro-4-iodobenzene

CAS Number: 136434-77-0
MDL Number: MFCD00042577
Molecular Formula: C6H3BrFI
Molecular Weight: 300.90
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 39
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 112 °C
Melting Pt: 39 °C
Flash Pt: 110 °C
MDL Number: MFCD00042577
CAS Number: 136434-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d06b71b61b6748fad455fbb75a5e2826057e244e&vid=8c529880cf45ade83737ab2ff8f18b6161ea11aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR