N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B3007-25G B3007-5G
TCB3007-25GEA 153.26 CAD
TCB3007-25G TCB3007-5G
N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-D-leucine hydrate

CAS Number: 16937-99-8
MDL Number: MFCD00038294
Molecular Formula: C11H21NO4
Molecular Weight: 231.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 88
Specific rotation [a]20/D: 25 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-leucine, Boc-D-Leu-OH

Formula: C₁₁H₂₁NO₄
Melting Pt: 88 °C
MDL Number: MFCD00038294
CAS Number: 16937-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06c2e73040a678469900793958d21a21fa80310a&vid=06c2e73040a678469900793958d21a21fa80310a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR