2-Bromo-4,6-difluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Bromo-3,5-difluoro-2-hydroxybenzene

B3004-1G B3004-5G
TCB3004-1GEA 137.46 CAD
TCB3004-1G TCB3004-5G
2-Bromo-4,6-difluorophenol ≥98.0% (by GC, titration analysis)
2-Bromo-4,6-difluorophenol

CAS Number: 98130-56-4
MDL Number: MFCD00070752
Molecular Formula: C6H3BrF2O
Molecular Weight: 208.99
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 53
Flash Point (°C): 106
Specific Gravity (20/20): 1.85

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Boiling Pt: 53 °C
Density: 1.85 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00070752
CAS Number: 98130-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f6221ea412b68247aaf5e1b85a01c563ec815e3&vid=4f6221ea412b68247aaf5e1b85a01c563ec815e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR