5-Bromo-2-chlorotoluene ≥98.0%
Supplier: TCI America
B3002-5G
B3002-25G
TCB3002-5GEA
57.83
CAD
TCB3002-5G
TCB3002-25G
5-Bromo-2-chlorotoluene ≥98.0%
5-Bromo-2-chlorotoluene
CAS Number: 54932-72-8
MDL Number: MFCD00018503
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 110
Specific Gravity (20/20): 1.56
Formula:
C₇H₆BrCl Boiling Pt: 93 °C Density: 1.56 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00018503 CAS Number: 54932-72-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...