N-Boc-D-valine ≥98.0%

Supplier: TCI America

Synonyms: Boc-D-Val-OH

B2991-5G B2991-25G
TCB2991-5GEA 37.92 CAD
TCB2991-5G TCB2991-25G
N-Boc-D-valine ≥98.0%
N-Boc-D-valine

CAS Number: 22838-58-0
MDL Number: MFCD00038282
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 79
Specific rotation [a]20/D: 6 deg (C=1, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-valine, Boc-D-Val-OH

Formula: C₁₀H₁₉NO₄
Melting Pt: 79 °C
MDL Number: MFCD00038282
CAS Number: 22838-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a998038eed564c2ce1d80b3bbd3a387007d7fa9&vid=4a998038eed564c2ce1d80b3bbd3a387007d7fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR