N-(tert-Butoxycarbonyl)-D-phenylalanine ≥98.0%
Supplier: TCI America
Synonyms:
N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH
B2989-25G
B2989-5G
TCB2989-25GEA
71.1
CAD
TCB2989-25G
TCB2989-5G
N-(tert-Butoxycarbonyl)-D-phenylalanine ≥98.0%
N-(tert-Butoxycarbonyl)-D-phenylalanine
CAS Number: 18942-49-9
MDL Number: MFCD00063149
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 87
Specific rotation [a]20/D: -25 deg (C=1, EtOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-phenylalanine, Boc-D-Phe-OH
Formula:
C₁₄H₁₉NO₄ Melting Pt: 87 °C |
MDL Number:
MFCD00063149 CAS Number: 18942-49-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...