Boc-D-Pro-OH ≥98.0%

Supplier: TCI America

Synonyms: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

B2977-25G B2977-5G
TCB2977-25GEA 109.02 CAD
TCB2977-25G TCB2977-5G
Boc-D-Pro-OH ≥98.0%
Boc-D-Pro-OH

CAS Number: 37784-17-1
MDL Number: MFCD00063226
Molecular Formula: C10H17NO4
Molecular Weight: 215.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 135
Specific rotation [a]20/D: 60 deg (C=2, AcOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-proline, Boc-D-Pro-OH

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Melting Pt: 135 °C
MDL Number: MFCD00063226
CAS Number: 37784-17-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db32a77a2c50f25ddd1f39b656dbf99f7fa0fd3d&vid=db32a77a2c50f25ddd1f39b656dbf99f7fa0fd3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR