N-(tert-Butoxycarbonyl)-D-methionine ≥95.0%
Supplier: TCI America
Synonyms:
(R)-2-((tert-Butoxycarbonyl)amino)-4-(methylthio)butanoic acid, Boc-D-Met-OH, N-Boc-D-methionine
B2967-25G
B2967-5G
TCB2967-25GEA
178.54
CAD
TCB2967-25G
TCB2967-5G
N-(tert-Butoxycarbonyl)-D-methionine ≥95.0%
N-(tert-Butoxycarbonyl)-D-methionine
CAS Number: 5241-66-7
MDL Number: MFCD00038256
Molecular Formula: C10H19NO4S
Molecular Weight: 249.33
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 22 deg (C=1, MeOH)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-D-methionine, Boc-D-Met-OH
Formula:
C₁₀H₁₉NO₄S MW: 249.33 g/mol |
MDL Number:
MFCD00038256 CAS Number: 5241-66-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...