3-Bromo-2-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-2-picoline

B2962-1G B2962-5G
TCB2962-1GEA 66.04 CAD
TCB2962-1G TCB2962-5G
3-Bromo-2-methylpyridine ≥98.0%
3-Bromo-2-methylpyridine

CAS Number: 38749-79-0
MDL Number: MFCD00191224
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 79
Specific Gravity (20/20): 1.52


Other Product Names / Synonyms: 3-Bromo-2-picoline

Formula: C₆H₆BrN
MW: 172.02
Boiling Pt: 76 °C
Density: 1.52 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00191224
CAS Number: 38749-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0586a4fc21fefd7f23f2e469bf757c15dbecd38&vid=e0586a4fc21fefd7f23f2e469bf757c15dbecd38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR