2-Bromo-4-chloro-1-fluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene

B2956-25G B2956-5G
TCB2956-25GEA 127.35 CAD
TCB2956-25G TCB2956-5G
2-Bromo-4-chloro-1-fluorobenzene ≥98.0%
2-Bromo-4-chloro-1-fluorobenzene

CAS Number: 1996-30-1
MDL Number: MFCD00672934
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 85
Specific Gravity (20/20): 1.74

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 90 °C
Density: 1.74 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00672934
CAS Number: 1996-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b32da8c5025beff9b75b4feb05638dba9698f1f3&vid=b32da8c5025beff9b75b4feb05638dba9698f1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR