4-Bromo-2-chloropyridine ≥98.0%

Supplier: TCI America

B2951-1G B2951-5G
TCB2951-1GEA 26.86 CAD
TCB2951-1G TCB2951-5G
4-Bromo-2-chloropyridine ≥98.0%
4-Bromo-2-chloropyridine

CAS Number: 73583-37-6
MDL Number: MFCD03840756
Molecular Formula: C5H3BrClN
Molecular Weight: 192.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 107
Freezing point (°C): 19

Formula: C₅H₃BrClN
MW: 192.44
Boiling Pt: 70 °C
Flash Pt: 107 °C
MDL Number: MFCD03840756
CAS Number: 73583-37-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8441d9c0a5c225ae3f53d8d4eb933bca720a45f9&vid=8441d9c0a5c225ae3f53d8d4eb933bca720a45f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR