4-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromo-2-fluorobenzotrifluoride, 1-Bromo-3-fluoro-4-(trifluoromethyl)benzene
B2946-5G
B2946-25G
TCB2946-5GEA
55.3
CAD
TCB2946-5G
TCB2946-25G
4-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
4-Bromo-α,α,α,2-tetrafluorotoluene
CAS Number: 142808-15-9
MDL Number: MFCD01075254
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.72
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Bromo-alpha,alpha,alpha,2-tetrafluorotoluene
Formula:
C₇H₃BrF₄ MW: 243 g/mol Density: 1.72 (20 °C) |
MDL Number:
MFCD01075254 CAS Number: 142808-15-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...