3-Bromothiophene-2-carbaldehyde ≥95.0%

Supplier: TCI America

Synonyms: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

B2934-1G B2934-25G B2934-5G
TCB2934-1GEA 53.72 CAD
TCB2934-1G TCB2934-25G TCB2934-5G
3-Bromothiophene-2-carbaldehyde ≥95.0%
3-Bromothiophene-2-carbaldehyde

CAS Number: 930-96-1
MDL Number: MFCD00126680
Molecular Formula: C5H3BrOS
Molecular Weight: 191.04
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.78


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Bromo-2-formylthiophene

Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 115 °C
Density: 1.78 (20 °C)
MDL Number: MFCD00126680
CAS Number: 930-96-1

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR