(S)-(-)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
CAS Number: 147081-44-5
MDL Number: MFCD03419271
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 91
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: -3 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3S)-(-)-1-Boc-3-aminopyrrolidine, (3S)-(-)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine, (3S)-(-)-3-Amino-1-Boc-pyrrolidine
Formula:
C₉H₁₈N₂O₂ MW: 186.25 g/mol Boiling Pt: 217 °C Density: 1.03 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD03419271 CAS Number: 147081-44-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...