(R)-(+)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine

B2920-5G B2920-1G
TCB2920-5GEA 262.28 CAD
TCB2920-5G TCB2920-1G
(R)-(+)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(R)-(+)-1-Boc-3-aminopyrrolidine

CAS Number: 147081-49-0
MDL Number: MFCD03427030
Molecular Formula: C9H18N2O2
Molecular Weight: 186.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 244
Flash Point (°C): 91
Specific Gravity (20/20): 1.03
Specific rotation [a]20/D: 4 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine

Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 244 °C
Density: 1.03 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD03419272
CAS Number: 147081-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdf2045f76b0cde94eeb7b54cc41294857c46ac8&vid=fdf2045f76b0cde94eeb7b54cc41294857c46ac8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR