(S)-2-Methylpropane-2-sulfinamide ≥98.0%
Supplier: TCI America
B2908-1G
B2908-5G
TCB2908-1GEA
80.58
CAD
TCB2908-1G
TCB2908-5G
(S)-2-Methylpropane-2-sulfinamide ≥98.0%
(S)-2-Methylpropane-2-sulfinamide
CAS Number: 343338-28-3
MDL Number: MFCD05861480
Molecular Formula: C4H11NOS
Molecular Weight: 121.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 106
Specific rotation [a]20/D: -4 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Sulfinic Acid Derivatives, Chiral Auxiliaries
Other Product Names / Synonyms: (S)-(-)-2-Methyl-2-propanesulfinamide
Formula:
C₄H₁₁NOS Melting Pt: 106 °C |
MDL Number:
MFCD05861480 CAS Number: 343338-28-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...