(R)-(+)-2-Methyl-2-propanesulfinamide ≥98.0%
Supplier: TCI America
Synonyms:
(R)-2-methylpropane-2-sulphinamide, (R)-2-methylpropane-2-sulfinamide, R)-(+)-tert-Butyl sulphinamide, (R)-2-Methyl-2-propanesulfinamide, (R)-(+)-tert-Butylsulfinamide, (R)-(+)-t-Butylsulfinamide, (R)-2-Methyl-2-propanesulphinamide, (R)-(+)-tert-Butylsulphinamide, R)-(+)-tert-Butyl sulfinamide, (R)-(+)-t-Butylsulphinamide
CAS Number: 196929-78-9
MDL Number: MFCD05861479
Molecular Formula: C4H11NOS
Molecular Weight: 121.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 106
Specific rotation [a]20/D: 4 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Sulfinic Acid Derivatives, Chiral Auxiliaries
Other Product Names / Synonyms: (R)-(+)-2-Methyl-2-propanesulfinamide
Formula:
C₄H₁₁NOS MW: 121.20 g/mol Melting Pt: 106 °C |
MDL Number:
MFCD05861479 CAS Number: 196929-78-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...