3-Bromophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B2890-1G
B2890-5G
B2890-25G
TCB2890-1GEA
53.72
CAD
TCB2890-1G
TCB2890-5G
TCB2890-25G
3-Bromophenylboronic acid (contains varying amounts of Anhydride)
3-Bromophenylboronic acid
CAS Number: 89598-96-9
MDL Number: MFCD00239386
Molecular Formula: C6H6BBrO2
Molecular Weight: 200.83
Form: Crystal
Color: White
Melting point (°C): 168
Other Product Names / Synonyms: 3-Bromobenzeneboronic Acid
Formula:
C₆H₆BBrO₂ Melting Pt: 168 °C |
MDL Number:
MFCD00239386 CAS Number: 89598-96-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...