2-Bromo-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2887-25G B2887-5G
TCB2887-25GEA 277.76 CAD
TCB2887-25G TCB2887-5G
2-Bromo-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-4-fluorobenzoic acid

CAS Number: 1006-41-3
MDL Number: MFCD00055370
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174

Formula: C₇H₄BrFO₂
Melting Pt: 174 °C
MDL Number: MFCD00055370
CAS Number: 1006-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0eb5c74e4014f8fc81315dd134b335787003a26&vid=9efc0381f95342ad3be05546468ff45c0984679a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR