5-Bromo-2-iodopyridine ≥97.0%

Supplier: TCI America

B2877-5G B2877-25G
TCB2877-5GEA 75.84 CAD
TCB2877-5G TCB2877-25G
5-Bromo-2-iodopyridine ≥97.0%
5-Bromo-2-iodopyridine

CAS Number: 223463-13-6
MDL Number: MFCD00691506
Molecular Formula: C5H3BrIN
Molecular Weight: 283.89
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 115

Formula: C₅H₃BrIN
Melting Pt: 115 °C
MDL Number: MFCD00691506
CAS Number: 223463-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17c0a1ea80cb90de5912069de5580766a63d1123&vid=17c0a1ea80cb90de5912069de5580766a63d1123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR