3-Bromocumene ≥98.0%

Supplier: TCI America

B2875-25G B2875-5G
TCB2875-25GEA 219.62 CAD
TCB2875-25G TCB2875-5G
3-Bromocumene ≥98.0%
3-Bromocumene

CAS Number: 5433-01-2
MDL Number: MFCD01318112
Molecular Formula: C9H11Br
Molecular Weight: 199.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Specific Gravity (20/20): 1.29


Other Product Names / Synonyms: 1-Bromo-3-isopropylbenzene

Formula: C₉H₁₁Br
Boiling Pt: 210 °C
Density: 1.29 (20 °C)
MDL Number: MFCD01318112
CAS Number: 5433-01-2
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7a16e5a88e59dbec91e8086f0b3fcd73238d118&vid=f7a16e5a88e59dbec91e8086f0b3fcd73238d118&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR