7-Bromoindole ≥97.0%

Supplier: TCI America

Synonyms: 7-Bromo-1H-indole

B2873-1G B2873-5G
TCB2873-1GEA 150.42 CAD
TCB2873-1G TCB2873-5G
7-Bromoindole ≥97.0%
7-Bromoindole

CAS Number: 51417-51-7
MDL Number: MFCD00799492
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 113

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 46 °C
Flash Pt: 113 °C
MDL Number: MFCD00799492
CAS Number: 51417-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abcc7de6b5a98ea1b21d44bd411462be30b154c2&vid=abcc7de6b5a98ea1b21d44bd411462be30b154c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR