5-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%

Supplier: TCI America

B2872-25G B2872-5G
TCB2872-25GEA 118.5 CAD
TCB2872-25G TCB2872-5G
5-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
5-Bromo-α,α,α,2-tetrafluorotoluene

CAS Number: 393-37-3
MDL Number: MFCD00042493
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 110
Specific Gravity (20/20): 1.73


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-alpha,alpha,alpha,2-tetrafluorotoluene, 4-Bromo-1-fluoro-2-(trifluoromethyl)benzene

Formula: C₇H₃BrF₄
Boiling Pt: 147 °C
Density: 1.73 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00042493
CAS Number: 393-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e357b82ff5dfc4a3ba88640668d034c80fe64195&vid=e357b82ff5dfc4a3ba88640668d034c80fe64195&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR