5-(tert-Butoxycarbonylamino)-1-pentanol ≥97.0%

Supplier: TCI America

Synonyms: N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(5-Hydroxypentyl)carbamate, tert-Butyl (5-hydroxypentyl)carbamate, N-Boc-5-aminopentanol, 5-(Boc-amino)-1-pentanol

B2869-5G B2869-25G
TCB2869-5GEA 247.9 CAD
TCB2869-5G TCB2869-25G
5-(tert-Butoxycarbonylamino)-1-pentanol ≥97.0%
5-(tert-Butoxycarbonylamino)-1-pentanol

CAS Number: 75178-90-4
Molecular Formula: C10H21NO3
Molecular Weight: 203.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 109
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: N-Boc-5-aminopentanol, tert-Butyl N-(5-Hydroxypentyl)carbamate, N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Density: 1.01 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD01862955
CAS Number: 75178-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3986957d1b759813a7585a53b35d39b737897c5&vid=e3986957d1b759813a7585a53b35d39b737897c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR