Bis[2-(diphenylphosphino)phenyl]ether ≥98.0%
Supplier: TCI America
Synonyms:
2,2'-Bis(diphenylphosphino)diphenyl Ether, DPEphos, Bis(2-diphenylphosphinophenyl)ether, (Oxydi-2,1-phenylene)bis(diphenylphosphine), (Oxybis(2,1-phenylene))bis(diphenylphosphine)
CAS Number: 166330-10-5
MDL Number: MFCD00233863
Molecular Formula: C36H28OP2
Molecular Weight: 538.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 186
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: (Oxydi-2,1-phenylene)bis(diphenylphosphine), 2,2'-Bis(diphenylphosphino)diphenyl Ether, DPEphos
Formula:
C₃₆H₂₈OP₂ MW: 538.57 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD00233863 CAS Number: 166330-10-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...