5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98.0%
Supplier: TCI America
B2853-25G
B2853-5G
TCB2853-25GEA
175.38
CAD
TCB2853-25G
TCB2853-5G
5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98.0%
5-Bromo-α,α,α-trifluoro-2-nitrotoluene
CAS Number: 344-38-7
MDL Number: MFCD00014706
Molecular Formula: C7H3BrF3NO2
Molecular Weight: 270.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 35
Flash Point (°C): 113
Other Product Names / Synonyms: 4-Bromo-1-nitro-2-(trifluoromethyl)benzene
Formula:
C₇H₃BrF₃NO₂ Boiling Pt: 100 °C Melting Pt: 35 °C Flash Pt: 113 °C |
MDL Number:
MFCD00014706 CAS Number: 344-38-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...