1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2851-25G B2851-5G
TCB2851-25GEA 551.01 CAD
TCB2851-25G TCB2851-5G
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulphonyl)imide

CAS Number: 223437-11-4
Molecular Formula: C11H20F6N2O4S2
Molecular Weight: 422.40
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.40

Formula: C₁₁H₂₀F₆N₂O₄S₂
Density: 1.40 (20 °C)
MDL Number: MFCD07784447
CAS Number: 223437-11-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR