5-Bromo-2-hydroxypyrimidine ≥95.0%
Supplier: TCI America
B2848-25G
B2848-5G
TCB2848-25GEA
178.54
CAD
TCB2848-25G
TCB2848-5G
5-Bromo-2-hydroxypyrimidine ≥95.0%
5-Bromo-2-hydroxypyrimidine
CAS Number: 38353-06-9
Molecular Formula: C4H3BrN2O
Molecular Weight: 174.99
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 235
Other Product Names / Synonyms: 5-Bromo-2-pyrimidinol, 5-Bromo-2(1H)-pyrimidinone
Formula:
C₄H₃BrN₂O Melting Pt: 235 °C |
MDL Number:
MFCD01318956 CAS Number: 38353-06-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...