5-Bromo-2-chloropyrimidine ≥97.0%

Supplier: TCI America

B2847-1G B2847-5G B2847-25G
TCB2847-1GEA 33.18 CAD
TCB2847-1G TCB2847-5G TCB2847-25G
5-Bromo-2-chloropyrimidine ≥97.0%
5-Bromo-2-chloropyrimidine

CAS Number: 32779-36-5
Molecular Formula: C4H2BrClN2
Molecular Weight: 193.43
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 78

Formula: C₄H₂BrClN₂
MW: 193.43 g/mol
Boiling Pt: 95 °C
Melting Pt: 78 °C
MDL Number: MFCD00483232
CAS Number: 32779-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ec732d9f2d218b4e73f046c728396e69903e3f&vid=f4ec732d9f2d218b4e73f046c728396e69903e3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR