4-Bromo-3,5-dimethoxybenzoic acid ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
B2838-25G
B2838-5G
TCB2838-25GEA
91.64
CAD
TCB2838-25G
TCB2838-5G
4-Bromo-3,5-dimethoxybenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Bromo-3,5-dimethoxybenzoic acid
CAS Number: 56518-42-4
MDL Number: MFCD01632140
Molecular Formula: C9H9BrO4
Molecular Weight: 261.07
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 252
Formula:
C₉H₉BrO₄ MW: 261.07 g/mol Melting Pt: 252 °C |
MDL Number:
MFCD01632140 CAS Number: 56518-42-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...