2-Bromopyrimidine ≥98.0%

Supplier: TCI America

B2832-5G B2832-25G
TCB2832-5GEA 151.68 CAD
TCB2832-5G TCB2832-25G
2-Bromopyrimidine ≥98.0%
2-Bromopyrimidine

CAS Number: 4595-60-2
MDL Number: MFCD00014601
Molecular Formula: C4H3BrN2
Molecular Weight: 158.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 64
Melting point (°C): 53
Storage Temperature: 0-10°C

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 64 °C
Melting Pt: 53 °C
MDL Number: MFCD00014601
CAS Number: 4595-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1a9a661c7816e03aa0f4535fdf5e9427eb2f084&vid=b1a9a661c7816e03aa0f4535fdf5e9427eb2f084&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR