3-Bromo-4-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-Bromo-4-picoline

B2831-5G B2831-1G B2831-25G
TCB2831-5GEA 53.72 CAD
TCB2831-5G TCB2831-1G TCB2831-25G
3-Bromo-4-methylpyridine ≥98.0%
3-Bromo-4-methylpyridine

CAS Number: 3430-22-6
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 76
Flash Point (°C): 79
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 3-Bromo-4-picoline

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 76 °C
Density: 1.53 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00082592
CAS Number: 3430-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=873e8e6c62eda8ec7f7da4de39dc70f7d317d1c1&vid=873e8e6c62eda8ec7f7da4de39dc70f7d317d1c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR