2-Bromo-4-nitroanisole ≥98.0%
Supplier: TCI America
B2830-25G
B2830-5G
TCB2830-25GEA
654.12
CAD
TCB2830-25G
TCB2830-5G
2-Bromo-4-nitroanisole ≥98.0%
2-Bromo-4-nitroanisole
CAS Number: 5197-28-4
Molecular Formula: C7H6BrNO3
Molecular Weight: 232.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 104
Other Product Names / Synonyms: 3-Bromo-4-methoxynitrobenzene
Formula:
C₇H₆BrNO₃ Melting Pt: 104 °C |
MDL Number:
MFCD00041242 CAS Number: 5197-28-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...