4-Bromothiophene-2-carbaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde
B2827-5G
B2827-25G
TCB2827-5GEA
66.36
CAD
TCB2827-5G
TCB2827-25G
4-Bromothiophene-2-carbaldehyde ≥98.0%
4-Bromothiophene-2-carbaldehyde
CAS Number: 18791-75-8
Molecular Formula: C5H3BrOS
Molecular Weight: 191.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 46
Flash Point (°C): 110
Other Product Names / Synonyms: 4-Bromo-2-formylthiophene
Formula:
C₅H₃BrOS MW: 191.05 g/mol Boiling Pt: 115 °C Melting Pt: 46 °C Flash Pt: 110 °C |
MDL Number:
MFCD00005431 CAS Number: 18791-75-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...